4-(Thiomorpholin-4-ylsulphonyl)benzeneboronic acid 96%

Supplier: BLD PHARMATECH GMBH

Synonyms: N-Thiomorpholin-4-yl 4-boronobenzenesulphonamide

BLDPBD221152-1GEA 185 EUR
BLDPBD221152-1G BLDPBD221152-100MG BLDPBD221152-250MG
4-(Thiomorpholin-4-ylsulphonyl)benzeneboronic acid 96%
4-(Thiomorpholin-4-ylsulphonyl)benzeneboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₀H₁₄BNO₄S₂
MW: 287,163 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07363757
CAS Number: 871329-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR