N(α)-Cbz-D-histidine 97%

Synonyms: Z-D-His-OH,N(α)-Benzyloxycarbonyl-D-histidine

BD22135-250MG
BLDPBD22135-250MGEA 10.5 EUR
BLDPBD22135-250MG
N(α)-Cbz-D-histidine 97%
N(α)-Cbz-D-histidine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₄H₁₅N₃O₄
MW: 289,3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00067702
CAS Number: 67424-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR