3-(Bromomethyl)pyridazine hydrobromide 97%
Supplier: BLD PHARMATECH GMBH
|
|
BLDPBD223837-1GEA
349
EUR
BLDPBD223837-1G
BLDPBD223837-250MG
BLDPBD223837-100MG
BLDPBD223837-5G
3-(Bromomethyl)pyridazine hydrobromide 97%
3-(Bromomethyl)pyridazine hydrobromide
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₅H₅BrN₂·BrH MW: 253,923 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD20526346 CAS Number: 1452483-94-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...