2,1,3-Benzothiadiazol-4-ylamine 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: Benzo[c][1,2,5]thiadiazol-4-amine4-Aminobenzo-2,1,3-thiadiazole

BLDPBD22551-1GEA 44.1 EUR
BLDPBD22551-1G BLDPBD22551-250MG BLDPBD22551-5G
2,1,3-Benzothiadiazol-4-ylamine 98%
2,1,3-Benzothiadiazol-4-ylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₅N₃S
MW: 151,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005810
CAS Number: 767-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR