1-Isobutyl-1H-pyrazole-4-boronic acid 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: (1-Isobutyl-1H-pyrazol-4-yl)boronic acid

BLDPBD226366-5GEA 398 EUR
BLDPBD226366-5G
1-Isobutyl-1H-pyrazole-4-boronic acid 98%
1-Isobutyl-1H-pyrazole-4-boronic acid

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formula: C₇H₁₃BN₂O₂
MW: 168,001 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09951916
CAS Number: 929094-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR