7-Quinolineboronic acid 98%
|
|
Synonyms:
7-Quinolinylboronic acid
, Quinoline-7-boronic acid
BLDPBD227137-250MGEA
69.3
EUR
BLDPBD227137-250MG
BLDPBD227137-1G
BLDPBD227137-100MG
BLDPBD227137-5G
7-Quinolineboronic acid 98%
7-Quinolineboronic acid
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₉H₈BNO₂ MW: 172,9800 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11100750 CAS Number: 629644-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...