3-Chloro-2-nitrophenol 95%
|
|
BLDPBD227189-250MGEA
24.2
EUR
BLDPBD227189-250MG
BLDPBD227189-5G
BLDPBD227189-10G
BLDPBD227189-25G
BLDPBD227189-1G
3-Chloro-2-nitrophenol 95%
3-Chloro-2-nitrophenol
Storage condition: Sealed in dry, Room Temperature
Formula:
C₆H₄ClNO₃ MW: 173,554 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08703192 CAS Number: 17802-02-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...