1-Methyl-1H-indazole-4-boronic acid pinacol ester 96%

Synonyms: 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

BLDPBD227620-1GEA 76.7 EUR
BLDPBD227620-1G BLDPBD227620-100MG BLDPBD227620-5G BLDPBD227620-250MG
1-Methyl-1H-indazole-4-boronic acid pinacol ester 96%
1-Methyl-1H-indazole-4-boronic acid pinacol ester

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₄H₁₉BN₂O₂
MW: 258,1239 g/mol
Storage Temperature: Freezer
MDL Number: MFCD10700158
CAS Number: 885698-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR