2,4,5-Trichloropyridine 95%

Supplier: BLD PHARMATECH GMBH

BLDPBD228466-250MGEA 9.5 EUR
BLDPBD228466-250MG BLDPBD228466-1G BLDPBD228466-5G BLDPBD228466-100MG
2,4,5-Trichloropyridine 95%
2,4,5-Trichloropyridine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅H₂Cl₃N
MW: 182,435 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09028037
CAS Number: 55934-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR