1,2,4,5-Tetrachloro-3,6-dimethoxybenzene 95%

BLDPBD228538-100MGEA 43.1 EUR
BLDPBD228538-100MG BLDPBD228538-1G BLDPBD228538-250MG
1,2,4,5-Tetrachloro-3,6-dimethoxybenzene 95%
1,2,4,5-Tetrachloro-3,6-dimethoxybenzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₆Cl₄O₂
MW: 275,94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22421619
CAS Number: 944-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR