1-Bromo-4,5-dichloro-2-nitrobenzene 97%
|
|
BLDPBD229500-25GEA
187
EUR
BLDPBD229500-25G
BLDPBD229500-250MG
BLDPBD229500-5G
BLDPBD229500-10G
BLDPBD229500-1G
1-Bromo-4,5-dichloro-2-nitrobenzene 97%
1-Bromo-4,5-dichloro-2-nitrobenzene
Storage condition: Sealed in dry, Room Temperature
Formula:
C₆H₂BrCl₂NO₂ MW: 270,9000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07780670 CAS Number: 93361-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...