1-Bromo-2-chloro-5-fluoro-4-nitrobenzene 97%

BLDPBD229759-250MGEA 14.7 EUR
BLDPBD229759-250MG BLDPBD229759-1G BLDPBD229759-10G BLDPBD229759-5G BLDPBD229759-25G BLDPBD229759-100G
1-Bromo-2-chloro-5-fluoro-4-nitrobenzene 97%
1-Bromo-2-chloro-5-fluoro-4-nitrobenzene

Storage condition: Sealed in dry, 2-8°C

Formula: C₆H₂BrClFNO₂
MW: 254,4400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11110551
CAS Number: 1027833-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR