2-Bromo-4-hydroxynicotinic acid 95%

Supplier: BLD PHARMATECH GMBH

Synonyms: 2-Bromo-3-carboxy-4-hydroxypyridine2-Bromo-4-hydroxypyridine-3-carboxylic acid

BLDPBD230366-5GEA 440 EUR
BLDPBD230366-5G BLDPBD230366-250MG BLDPBD230366-1G
2-Bromo-4-hydroxynicotinic acid 95%
2-Bromo-4-hydroxynicotinic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₆H₄BrNO₃
MW: 218,005 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12026106
CAS Number: 1150561-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR