3,4-Dihydro-2H-pyran-6-boronic acid, pinacol ester 95% stabilised

Synonyms: 2-(3,4-Dihydro-2H-pyran-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Stabilisation: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 1%

BLDPBD230722-100MGEA 14.7 EUR
BLDPBD230722-100MG BLDPBD230722-250MG
3,4-Dihydro-2H-pyran-6-boronic acid, pinacol ester 95% stabilised
3,4-Dihydro-2H-pyran-6-boronic acid, pinacol ester

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₁H₁₉BO₃
MW: 210,078 g/mol
Storage Temperature: Freezer
MDL Number: MFCD07368233
CAS Number: 1025707-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR