2-Bromo-4-chloro-1-(difluoromethoxy)benzene 97%

BD231008-1G BD231008-250MG BD231008-5G
BLDPBD231008-1GEA 26.3 EUR
BLDPBD231008-1G BLDPBD231008-250MG BLDPBD231008-5G
2-Bromo-4-chloro-1-(difluoromethoxy)benzene 97%
2-Bromo-4-chloro-1-(difluoromethoxy)benzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₄BrClF₂O
MW: 257,45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847190
CAS Number: 1214348-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR