4-Carboxy-3-chlorophenylboronic acid pinacol ester 96%
|
|
Synonyms:
4-Carboxy-3-chlorophenylboronic acid, pinacol ester
, 2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
BLDPBD231242-250MGEA
68.3
EUR
BLDPBD231242-250MG
4-Carboxy-3-chlorophenylboronic acid pinacol ester 96%
4-Carboxy-3-chlorophenylboronic acid pinacol ester
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₃H₁₆BClO₄ MW: 282,5277 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12546621 CAS Number: 890839-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...