2-Chloro-6-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile 98%

BD231321-5G BD231321-1G BD231321-250MG
BLDPBD231321-5GEA 506 EUR
BLDPBD231321-5G BLDPBD231321-1G BLDPBD231321-250MG
2-Chloro-6-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile 98%
2-Chloro-6-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₂H₁₃BClNO₂
MW: 249,50 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08669560
CAS Number: 883899-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR