8-Fluoro-2-quinolinol 97%

Synonyms: 8-Fluoroquinolin-2(1H)-one

BD231362-250MG BD231362-100MG BD231362-1G BD231362-5G BD231362-25G
BLDPBD231362-250MGEA 47.3 EUR
BLDPBD231362-250MG BLDPBD231362-100MG BLDPBD231362-1G BLDPBD231362-5G BLDPBD231362-25G
8-Fluoro-2-quinolinol 97%
8-Fluoro-2-quinolinol

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₆FNO
MW: 163,1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026113
CAS Number: 71738-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR