4-Bromo-2,3-dihydro-1H-inden-1-ol 95%

BLDPBD231747-5GEA 202 EUR
BLDPBD231747-5G BLDPBD231747-250MG BLDPBD231747-1G
4-Bromo-2,3-dihydro-1H-inden-1-ol 95%
4-Bromo-2,3-dihydro-1H-inden-1-ol

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₉BrO
MW: 213,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11219497
CAS Number: 16657-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR