4'-Chloro-2'-nitroacetophenone 98%

Synonyms: 1-(4-Chloro-2-nitrophenyl)ethanone

BLDPBD232246-1GEA 79.8 EUR
BLDPBD232246-1G BLDPBD232246-100MG BLDPBD232246-5G BLDPBD232246-250MG BLDPBD232246-10G BLDPBD232246-25G
4'-Chloro-2'-nitroacetophenone 98%
4'-Chloro-2'-nitroacetophenone

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₆ClNO₃
MW: 199,5900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09031331
CAS Number: 23082-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR