2'-Fluoro-3'-nitroacetophenone 98%

Synonyms: 1-(2-Fluoro-3-nitrophenyl)ethanone

BLDPBD232286-250MGEA 72.5 EUR
BLDPBD232286-250MG BLDPBD232286-1G BLDPBD232286-5G
2'-Fluoro-3'-nitroacetophenone 98%
2'-Fluoro-3'-nitroacetophenone

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₆FNO₃
MW: 183,1400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13194361
CAS Number: 873697-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR