(1H-Indazol-4-yl)acetic acid 95%

Synonyms: 4-(Carboxymethyl)-1H-indazole2-(1H-Indazol-4-yl)ethanoic acid

BLDPBD232351-100MGEA 216 EUR
BLDPBD232351-100MG BLDPBD232351-250MG
(1H-Indazol-4-yl)acetic acid 95%
(1H-Indazol-4-yl)acetic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₉H₈N₂O₂
MW: 176,172 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13183061
CAS Number: 902131-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR