4-(2-Morpholinoethyl)phenylboronic acid, pinacol ester 95%

Synonyms: 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenethyl)morpholine

BLDPBD232422-100MGEA 41 EUR
BLDPBD232422-100MG BLDPBD232422-250MG BLDPBD232422-1G
4-(2-Morpholinoethyl)phenylboronic acid, pinacol ester 95%
4-(2-Morpholinoethyl)phenylboronic acid, pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₈H₂₈BNO₃
MW: 317,231 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04440855
CAS Number: 364794-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR