3-Bromo-4-methylanisole 97%

Synonyms: 2-Bromo-4-methoxy-1-methylbenzene

BD232616-5G BD232616-500G BD232616-10G BD232616-100G BD232616-25G BD232616-1G
BLDPBD232616-5GEA 18.9 EUR
BLDPBD232616-5G BLDPBD232616-500G BLDPBD232616-10G BLDPBD232616-100G BLDPBD232616-25G BLDPBD232616-1G
3-Bromo-4-methylanisole 97%
3-Bromo-4-methylanisole

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₉BrO
MW: 201,0600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547797
CAS Number: 36942-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR