Benzo[1,2-b:3,4-b':5,6-b'']trithiophene 96%

BLDPBD232675-1GEA 907 EUR
BLDPBD232675-1G BLDPBD232675-100MG BLDPBD232675-250MG BLDPBD232675-50MG
Benzo[1,2-b:3,4-b':5,6-b'']trithiophene 96%
Benzo[1,2-b:3,4-b':5,6-b'']trithiophene

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₂H₆S₃
MW: 246,37 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22381057
CAS Number: 29150-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR