6-Bromoisatoic anhydride 95%

Supplier: BLD PHARMATECH GMBH

Synonyms: 5-Bromo-1H-benzo[d][1,3]oxazine-2,4-dione

BLDPBD232954-25GEA 289 EUR
BLDPBD232954-25G BLDPBD232954-250MG BLDPBD232954-5G BLDPBD232954-10G BLDPBD232954-1G
6-Bromoisatoic anhydride 95%
6-Bromoisatoic anhydride

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₈H₄BrNO₃
MW: 242,026 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15526667
CAS Number: 77603-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR