2-Bromo-5-(tert-butyl)aniline 98%

BD233079-100G BD233079-25G BD233079-1G BD233079-5G
BLDPBD233079-100GEA 741 EUR
BLDPBD233079-100G BLDPBD233079-25G BLDPBD233079-1G BLDPBD233079-5G
2-Bromo-5-(tert-butyl)aniline 98%
2-Bromo-5-(tert-butyl)aniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₀H₁₄BrN
MW: 228,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846050
CAS Number: 58164-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR