5-Chloro-6-oxo-1,6-dihydropyrimidine-4-carboxylic acid 95+%

BD233117-25G BD233117-1G BD233117-5G
BLDPBD233117-25GEA 698 EUR
BLDPBD233117-25G BLDPBD233117-1G BLDPBD233117-5G
5-Chloro-6-oxo-1,6-dihydropyrimidine-4-carboxylic acid 95+%
5-Chloro-6-oxo-1,6-dihydropyrimidine-4-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₅H₃ClN₂O₃
MW: 174,54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12911627
CAS Number: 914916-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR