3,5,6-Trichloropyridine-2,4-diamine 95%

BD233383-100MG BD233383-250MG BD233383-1G
BLDPBD233383-100MGEA 102 EUR
BLDPBD233383-100MG BLDPBD233383-250MG BLDPBD233383-1G
3,5,6-Trichloropyridine-2,4-diamine 95%
3,5,6-Trichloropyridine-2,4-diamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₅H₄Cl₃N₃
MW: 212,46 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00795936
CAS Number: 725208-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR