(11bS)-2,6-Bis(3,5-dimethylphenyl)-4-hydroxy-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 98%

BD233613-250MG BD233613-1G BD233613-100MG
BLDPBD233613-250MGEA 239 EUR
BLDPBD233613-250MG BLDPBD233613-1G BLDPBD233613-100MG
(11bS)-2,6-Bis(3,5-dimethylphenyl)-4-hydroxy-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 98%
(11bS)-2,6-Bis(3,5-dimethylphenyl)-4-hydroxy-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

Storage condition: Inert atmosphere, 2-8°C

Formula: C₃₆H₃₇O₄P
MW: 564,65 g/mol
Storage Temperature: Refrigerator
CAS Number: 2757287-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR