(R)-3,3'-Diiodo-2,2'-bis(methoxymethoxy)-1,1'-binaphthalene 98%

BD233632-1G BD233632-100MG BD233632-250MG
BLDPBD233632-1GEA 118 EUR
BLDPBD233632-1G BLDPBD233632-100MG BLDPBD233632-250MG
(R)-3,3'-Diiodo-2,2'-bis(methoxymethoxy)-1,1'-binaphthalene 98%
(R)-3,3'-Diiodo-2,2'-bis(methoxymethoxy)-1,1'-binaphthalene

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₂₄H₂₀I₂O₄
MW: 626,22 g/mol
Storage Temperature: Refrigerator
CAS Number: 189518-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR