(11bS)-4-Hydroxy-2,6-diphenyldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 97%

BLDPBD233678-100MGEA 101 EUR
BLDPBD233678-100MG BLDPBD233678-1G BLDPBD233678-250MG
(11bS)-4-Hydroxy-2,6-diphenyldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 97%
(11bS)-4-Hydroxy-2,6-diphenyldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

Storage condition: Inert atmosphere, 2-8°C

Formula: C₃₂H₂₁O₄P
MW: 500,48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17676194
CAS Number: 874948-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR