(11bS)-4-Hydroxy-2,6-bis(triphenylsilyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 98%

BLDPBD233686-100MGEA 62 EUR
BLDPBD233686-100MG BLDPBD233686-1G BLDPBD233686-250MG
(11bS)-4-Hydroxy-2,6-bis(triphenylsilyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 98%
(11bS)-4-Hydroxy-2,6-bis(triphenylsilyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₅₆H₄₁O₄PSi₂
MW: 865,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09265079
CAS Number: 929097-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR