Methyl 6-amino-1H-indazole-4-carboxylate 95%

BLDPBD233700-250MGEA 159 EUR
BLDPBD233700-250MG BLDPBD233700-1G BLDPBD233700-100MG
Methyl 6-amino-1H-indazole-4-carboxylate 95%
Methyl 6-amino-1H-indazole-4-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₉N₃O₂
MW: 191,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781325
CAS Number: 885518-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR