3-Bromo-6-chloro-4-nitro-1H-indazole 95%

BD233721-100MG BD233721-250MG
BLDPBD233721-100MGEA 33.6 EUR
BLDPBD233721-100MG BLDPBD233721-250MG
3-Bromo-6-chloro-4-nitro-1H-indazole 95%
3-Bromo-6-chloro-4-nitro-1H-indazole

Storage condition: Inert atmosphere, 2-8°C

Formula: C₇H₃BrClN₃O₂
MW: 276,48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07781689
CAS Number: 885519-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR