2-[2-(Vinyloxy)ethoxy]isoindoline-1,3-dione 95% mix stabilised

Supplier: BLD PHARMATECH GMBH

Synonyms: 2-(2-(Vinyloxy)ethoxy)isoindoline-1,3-dione

Stabilisation: Stabilised with 4-tert-butylpyrocatechol (TBC)

BLDPBD234373-100MGEA 42 EUR
BLDPBD234373-100MG BLDPBD234373-250MG BLDPBD234373-1G BLDPBD234373-5G
2-[2-(Vinyloxy)ethoxy]isoindoline-1,3-dione 95% mix stabilised
2-[2-(Vinyloxy)ethoxy]isoindoline-1,3-dione

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₂H₁₁NO₄
MW: 233,22 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15142583
CAS Number: 391212-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR