Bis(4-methoxyphenyl)phosphine oxide 95%

BD235632-5G BD235632-1G BD235632-250MG BD235632-25G
BLDPBD235632-5GEA 178 EUR
BLDPBD235632-5G BLDPBD235632-1G BLDPBD235632-250MG BLDPBD235632-25G
Bis(4-methoxyphenyl)phosphine oxide 95%
Bis(4-methoxyphenyl)phosphine oxide

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₄H₁₅O₃P
MW: 262,24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06754095
CAS Number: 15754-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR