1-Bromo-4-(trimethylsilyl)benzene 97%
|
|
Synonyms:
(4-Bromophenyl)trimethylsilane
BLDPBD236893-10GEA
62
EUR
BLDPBD236893-10G
BLDPBD236893-25G
BLDPBD236893-1G
BLDPBD236893-5G
BLDPBD236893-100G
1-Bromo-4-(trimethylsilyl)benzene 97%
1-Bromo-4-(trimethylsilyl)benzene
Storage condition: Sealed in dry, Room Temperature
Formula:
C₉H₁₃BrSi MW: 229,1890 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00094165 CAS Number: 6999-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...