3-Amino-5-chloropyrazine-2-carbonitrile 90%

BLDPBD237063-100MGEA 94.5 EUR
BLDPBD237063-100MG BLDPBD237063-250MG
3-Amino-5-chloropyrazine-2-carbonitrile 90%
3-Amino-5-chloropyrazine-2-carbonitrile

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₅H₃ClN₄
MW: 154,557 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10697779
CAS Number: 54632-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR