5-(4-Methylpiperazin-1-yl)-1H-indole 95%

BLDPBD237318-100MGEA 92.4 EUR
BLDPBD237318-100MG BLDPBD237318-250MG
5-(4-Methylpiperazin-1-yl)-1H-indole 95%
5-(4-Methylpiperazin-1-yl)-1H-indole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₃H₁₇N₃
MW: 215,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17392869
CAS Number: 412049-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR