2-Bromo-1-(1-methyl-1H-pyrazol-4-yl)ethanone 95%

BD238011-250MG
BLDPBD238011-250MGEA 223 EUR
BLDPBD238011-250MG
2-Bromo-1-(1-methyl-1H-pyrazol-4-yl)ethanone 95%
2-Bromo-1-(1-methyl-1H-pyrazol-4-yl)ethanone

Storage condition: Inert atmosphere, 2-8°C

Formula: C₆H₇BrN₂O
MW: 203,03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11182388
CAS Number: 706819-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR