7-Chloro-1H-indole-2-carbaldehyde 98%

BD238097-1G BD238097-250MG BD238097-100MG
BLDPBD238097-1GEA 991 EUR
BLDPBD238097-1G BLDPBD238097-250MG BLDPBD238097-100MG
7-Chloro-1H-indole-2-carbaldehyde 98%
7-Chloro-1H-indole-2-carbaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₆ClNO
MW: 179,60 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11040768
CAS Number: 53590-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR