1-(4-Bromo-3-fluorophenyl)-2-chloroethanone 97%

BD238253-100MG BD238253-1G BD238253-250MG
BLDPBD238253-100MGEA 120 EUR
BLDPBD238253-100MG BLDPBD238253-1G BLDPBD238253-250MG
1-(4-Bromo-3-fluorophenyl)-2-chloroethanone 97%
1-(4-Bromo-3-fluorophenyl)-2-chloroethanone

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₅BrClFO
MW: 251,48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11847747
CAS Number: 1260857-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR