6-(Benzyloxy)-4-fluoro-1H-indazole 97%

BLDPBD238313-100MGEA 120 EUR
BLDPBD238313-100MG BLDPBD238313-250MG BLDPBD238313-1G
6-(Benzyloxy)-4-fluoro-1H-indazole 97%
6-(Benzyloxy)-4-fluoro-1H-indazole

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₄H₁₁FN₂O
MW: 242,25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19443332
CAS Number: 1253792-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR