4-Bromo-1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%

BD238576-250MG BD238576-100MG BD238576-1G BD238576-5G
BLDPBD238576-250MGEA 20 EUR
BLDPBD238576-250MG BLDPBD238576-100MG BLDPBD238576-1G BLDPBD238576-5G
4-Bromo-1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%
4-Bromo-1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₀H₁₆BBrN₂O₂
MW: 286,96 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12405502
CAS Number: 942070-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR