2-Formylnicotinic acid 95+%

BLDPBD238615-1GEA 703 EUR
BLDPBD238615-1G BLDPBD238615-100MG BLDPBD238615-250MG
2-Formylnicotinic acid 95+%
2-Formylnicotinic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₇H₅NO₃
MW: 151,11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20642387
CAS Number: 23590-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR