4-Chloro-3-((3-chloro-5-(trifluoromethyl)pyridin-2-yl)oxy)aniline 95%

BD238696-250MG BD238696-1G BD238696-5G
BLDPBD238696-250MGEA 39.9 EUR
BLDPBD238696-250MG BLDPBD238696-1G BLDPBD238696-5G
4-Chloro-3-((3-chloro-5-(trifluoromethyl)pyridin-2-yl)oxy)aniline 95%
4-Chloro-3-((3-chloro-5-(trifluoromethyl)pyridin-2-yl)oxy)aniline

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₁₂H₇Cl₂F₃N₂O
MW: 323,10 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22200381
CAS Number: 87170-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR