(S)-4,5,6,7-Tetrahydro-1H-indazol-5-amine hydrochloride 97%

BLDPBD238708-1GEA 895 EUR
BLDPBD238708-1G BLDPBD238708-100MG BLDPBD238708-250MG
(S)-4,5,6,7-Tetrahydro-1H-indazol-5-amine hydrochloride 97%
(S)-4,5,6,7-Tetrahydro-1H-indazol-5-amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₁₂ClN₃
MW: 173,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11053505
CAS Number: 1263078-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR