Tetraphenylphosphonium phenolate 99% + mix

BLDPBD238895-10GEA 18.9 EUR
BLDPBD238895-10G BLDPBD238895-25G
Tetraphenylphosphonium phenolate 99% + mix
Tetraphenylphosphonium phenolate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₃₀H₂₅OP
MW: 432,49 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22200717
CAS Number: 15464-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR