2,6-Diiodo-4-(trifluoromethyl)aniline 95%

BD238984-250MG BD238984-1G BD238984-5G
BLDPBD238984-250MGEA 33.6 EUR
BLDPBD238984-250MG BLDPBD238984-1G BLDPBD238984-5G
2,6-Diiodo-4-(trifluoromethyl)aniline 95%
2,6-Diiodo-4-(trifluoromethyl)aniline

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₇H₄F₃I₂N
MW: 412,92 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11847372
CAS Number: 214400-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR